COM448686807 Combi-Blocks, Inc.

Namiki bbv-32465395

  • MDL number MFCD14586856
  • Cas Number : 99233-38-2
  • Molecular Weight : 153.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1804

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1804
Availability details for
Quantity
  • Availability
  • Add to Cart