COM448686808 Combi-Blocks, Inc.

methyl 3-methoxy-5-[4-(methoxycarbonyl)phenyl]benzoate

  • MDL number MFCD27578499
  • Cas Number : 1820618-09-4
  • Molecular Weight : 300.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1805

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1805
Availability details for
Quantity
  • Availability
  • Add to Cart