COM448686809 Combi-Blocks, Inc.

Methyl 5-bromo-2-butyramidobenzoate

  • MDL number MFCD02349188
  • Cas Number : 452349-54-1
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1807
Availability details for
Quantity
  • Availability
  • Add to Cart