COM448686816 Combi-Blocks, Inc.

dicyclohexylamine 1,4-dimethyl (2S)-2-{[(tert-butoxy)carbonyl]amino}butanedioate

  • MDL number MFCD27578502
  • Molecular Weight : 442.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1815

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1815
Availability details for
Quantity
  • Availability
  • Add to Cart