COM448686824 Combi-Blocks, Inc.

Methyl 3,5-difluoroisonicotinate

  • MDL number MFCD14698040
  • Cas Number : 1214324-60-3
  • Molecular Weight : 173.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1827

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1827
Availability details for
Quantity
  • Availability
  • Add to Cart