COM448686827 Combi-Blocks, Inc.

Methyl 1-(4,6-dimethylpyrimidin-2-yl)piperidine-4-carboxylate

  • MDL number MFCD10665022
  • Cas Number : 1090361-66-2
  • Molecular Weight : 249.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1832
Availability details for
Quantity
  • Availability
  • Add to Cart