COM448686829 Combi-Blocks, Inc.

methyl 3-(2,5-dimethoxyphenyl)benzoate

  • MDL number MFCD27578505
  • Cas Number : 1820675-12-4
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1834

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1834
Availability details for
Quantity
  • Availability
  • Add to Cart