COM448686830 Combi-Blocks, Inc.

methyl 3-{[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamoyl}propanoate

  • MDL number MFCD27578506
  • Molecular Weight : 333.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1835

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1835
Availability details for
Quantity
  • Availability
  • Add to Cart