COM448686832 Combi-Blocks, Inc.

Methyl 5-methoxy-[1,1'-biphenyl]-3-carboxylate

  • MDL number MFCD17214994
  • Cas Number : 184095-59-8
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1840

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1840
Availability details for
Quantity
  • Availability
  • Add to Cart