COM448686835 Combi-Blocks, Inc.

methyl 3-{[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamoyl}propanoate

  • MDL number MFCD27578508
  • Molecular Weight : 333.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1848

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1848
Availability details for
Quantity
  • Availability
  • Add to Cart