COM448686838 Combi-Blocks, Inc.

methyl 2-fluoro-4-[3-(2-methoxy-2-oxoethyl)phenyl]benzoate

  • MDL number MFCD27578510
  • Cas Number : 1820711-15-6
  • Molecular Weight : 302.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1852

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1852
Availability details for
Quantity
  • Availability
  • Add to Cart