COM448686840 Combi-Blocks, Inc.

methyl 4-[4-(benzyloxy)phenyl]-3-methoxybenzoate

  • MDL number MFCD27578511
  • Cas Number : 1820641-68-6
  • Molecular Weight : 348.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1854
Availability details for
Quantity
  • Availability
  • Add to Cart