COM448686843 Combi-Blocks, Inc.

methyl (2E)-3-[6-(trifluoromethyl)pyridin-3-yl]prop-2-enoate

  • MDL number MFCD27578513
  • Cas Number : 1119807-17-8
  • Molecular Weight : 231.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1857

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1857
Availability details for
Quantity
  • Availability
  • Add to Cart