COM448686844 Combi-Blocks, Inc.

methyl 3-(4-formylphenyl)-5-(trifluoromethyl)benzoate

  • MDL number MFCD27578514
  • Cas Number : 1820649-85-1
  • Molecular Weight : 308.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1858

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1858
Availability details for
Quantity
  • Availability
  • Add to Cart