COM448686845 Combi-Blocks, Inc.

methyl 4-(2-methoxy-5-methylphenyl)benzoate

  • MDL number MFCD27578515
  • Cas Number : 1820618-07-2
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1860

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1860
Availability details for
Quantity
  • Availability
  • Add to Cart