COM448686846 Combi-Blocks, Inc.

Methyl 2'-(benzyloxy)[1,1'-biphenyl]-3-carboxylate

  • MDL number MFCD06802262
  • Cas Number : 893736-43-1
  • Molecular Weight : 318.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1863

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1863
Availability details for
Quantity
  • Availability
  • Add to Cart