COM448686847 Combi-Blocks, Inc.

1-(9H-Fluoren-9-ylmethyl) 4-methyl piperidine-1,4-dicarboxylate

  • MDL number MFCD18328518
  • Cas Number : 1340322-21-5
  • Molecular Weight : 365.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1864

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1864
Availability details for
Quantity
  • Availability
  • Add to Cart