COM448686855 Combi-Blocks, Inc.

methyl 1-[(4-methylbenzene)sulfonyl]azetidine-3-carboxylate

  • MDL number MFCD27578520
  • Cas Number : 1820684-87-4
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1877

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1877
Availability details for
Quantity
  • Availability
  • Add to Cart