COM448686860 Combi-Blocks, Inc.

methyl 1-hydroxy-2-phenyl-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD27578525
  • Molecular Weight : 268.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1882

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1882
Availability details for
Quantity
  • Availability
  • Add to Cart