COM448686864 Combi-Blocks, Inc.

1,2-dimethyl 4-(2,5-dioxopyrrol-1-yl)phthalate

  • MDL number MFCD27578529
  • Cas Number : 62351-82-0
  • Molecular Weight : 289.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1886

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1886
Availability details for
Quantity
  • Availability
  • Add to Cart