COM448686866 Combi-Blocks, Inc.

4-(4-(Trifluoromethoxy)phenyl)picolinic acid methyl ester

  • MDL number MFCD18408455
  • Cas Number : 1261617-85-9
  • Molecular Weight : 297.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1888
Availability details for
Quantity
  • Availability
  • Add to Cart