COM448686873 Combi-Blocks, Inc.

methyl 2-amino-2,3,3-trimethylbutanoate hydrochloride

  • MDL number MFCD27578533
  • Cas Number : 1820741-04-5
  • Molecular Weight : 195.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1898
Availability details for
Quantity
  • Availability
  • Add to Cart