COM448686879 Combi-Blocks, Inc.

methyl 1-isopropyl-2-(trifluoromethyl)-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD27578538
  • Cas Number : 1390621-32-5
  • Molecular Weight : 286.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1906

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1906
Availability details for
Quantity
  • Availability
  • Add to Cart