COM448686881 Combi-Blocks, Inc.

methyl 3-(3,5-difluorophenyl)-5-(trifluoromethyl)benzoate

  • MDL number MFCD27578540
  • Cas Number : 1820707-76-3
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1908

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1908
Availability details for
Quantity
  • Availability
  • Add to Cart