COM448686884 Combi-Blocks, Inc.

methyl 5-[3-(methoxycarbonyl)phenyl]pyridine-3-carboxylate

  • MDL number MFCD27578542
  • Cas Number : 1820665-97-1
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1911

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1911
Availability details for
Quantity
  • Availability
  • Add to Cart