COM448686885 Combi-Blocks, Inc.

Methyl 3'-fluoro[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD05979259
  • Cas Number : 80254-86-0
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1912

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1912
Availability details for
Quantity
  • Availability
  • Add to Cart