COM448686886 Combi-Blocks, Inc.

methyl 6-(4-chlorophenyl)pyridine-3-carboxylate

  • MDL number MFCD27578543
  • Cas Number : 149467-80-1
  • Molecular Weight : 247.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1914

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1914
Availability details for
Quantity
  • Availability
  • Add to Cart