COM448686888 Combi-Blocks, Inc.

Methyl 2-(4-chlorophenyl)benzoate

  • MDL number MFCD05979380
  • Cas Number : 17103-28-5
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1918
Availability details for
Quantity
  • Availability
  • Add to Cart