COM448686890 Combi-Blocks, Inc.

Methyl 1-[(5-bromothiophen-2-yl)sulfonyl]piperidine-3-carboxylate

  • MDL number MFCD25564639
  • Cas Number : 1557659-17-2
  • Molecular Weight : 368.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1920
Availability details for
Quantity
  • Availability
  • Add to Cart