COM448686894 Combi-Blocks, Inc.

methyl 2-[4-(3,5-difluorophenyl)phenyl]acetate

  • MDL number MFCD27578547
  • Cas Number : 1820716-60-6
  • Molecular Weight : 262.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1926

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1926
Availability details for
Quantity
  • Availability
  • Add to Cart