COM448686896 Combi-Blocks, Inc.

methyl 3-fluoro-5-(5-fluoro-2-methylphenyl)benzoate

  • MDL number MFCD27578548
  • Cas Number : 1820675-13-5
  • Molecular Weight : 262.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1929

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1929
Availability details for
Quantity
  • Availability
  • Add to Cart