COM448686898 Combi-Blocks, Inc.

Methyl 4'-chloro[1,1'-biphenyl]-3-carboxylate

  • MDL number MFCD05978894
  • Cas Number : 773134-21-7
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1931

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1931
Availability details for
Quantity
  • Availability
  • Add to Cart