COM448686907 Combi-Blocks, Inc.

methyl 3-[4-(methoxycarbonyl)phenyl]-5-nitrobenzoate

  • MDL number MFCD27578556
  • Cas Number : 1820683-62-2
  • Molecular Weight : 315.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1941

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1941
Availability details for
Quantity
  • Availability
  • Add to Cart