COM448686908 Combi-Blocks, Inc.

methyl 3-chloro-5-(3,4-dichlorophenyl)benzoate

  • MDL number MFCD27578557
  • Cas Number : 1820648-38-1
  • Molecular Weight : 315.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1943

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1943
Availability details for
Quantity
  • Availability
  • Add to Cart