COM448686910 Combi-Blocks, Inc.

Benzoic acid, 3-borono-5-[[(1,1-dimethylethoxy)carbonyl]amino]-, 1-methyl ester

  • MDL number MFCD22571996
  • Cas Number : 428820-82-0
  • Molecular Weight : 295.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1945

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1945
Availability details for
Quantity
  • Availability
  • Add to Cart