COM448686913 Combi-Blocks, Inc.

methyl (2S)-6-{[(tert-butoxy)carbonyl]amino}-2-acetamidohexanoate

  • MDL number MFCD27578559
  • Cas Number : 16679-49-5
  • Molecular Weight : 302.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1948

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1948
Availability details for
Quantity
  • Availability
  • Add to Cart