COM448686916 Combi-Blocks, Inc.

methyl 2-chloro-4-[3-(methoxycarbonyl)phenyl]benzoate

  • MDL number MFCD27578561
  • Cas Number : 1820683-22-4
  • Molecular Weight : 304.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1954

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1954
Availability details for
Quantity
  • Availability
  • Add to Cart