COM448686917 Combi-Blocks, Inc.

methyl 4-chloro-3-phenylbenzoate

  • MDL number MFCD27578562
  • Cas Number : 1082720-85-1
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1956

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1956
Availability details for
Quantity
  • Availability
  • Add to Cart