COM448686919 Combi-Blocks, Inc.

methyl 4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate

  • MDL number MFCD27578563
  • Cas Number : 1820673-41-3
  • Molecular Weight : 271.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1959
Availability details for
Quantity
  • Availability
  • Add to Cart