COM448686921 Combi-Blocks, Inc.

methyl 3-(4-methoxy-2-methylphenyl)-2-methylbenzoate

  • MDL number MFCD27578565
  • Cas Number : 1820618-12-9
  • Molecular Weight : 270.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1963

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1963
Availability details for
Quantity
  • Availability
  • Add to Cart