COM448686922 Combi-Blocks, Inc.

(2E)-3-[1,1'-Biphenyl]-4-yl-2-propenoic acid, methyl ester

  • MDL number MFCD22124787
  • Cas Number : 22837-75-8
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1966
Availability details for
Quantity
  • Availability
  • Add to Cart