COM448686924 Combi-Blocks, Inc.

methyl 3-(naphthalen-1-yl)-5-nitrobenzoate

  • MDL number MFCD27578567
  • Cas Number : 1820642-15-6
  • Molecular Weight : 307.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1969

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1969
Availability details for
Quantity
  • Availability
  • Add to Cart