COM448686927 Combi-Blocks, Inc.

methyl 5-(3,5-dichlorophenyl)pyridine-2-carboxylate

  • MDL number MFCD25440597
  • Cas Number : 1361889-13-5
  • Molecular Weight : 282.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1973

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1973
Availability details for
Quantity
  • Availability
  • Add to Cart