COM448686928 Combi-Blocks, Inc.

Methyl 1-[(5-bromothiophen-2-yl)sulfonyl]pyrrolidine-2-carboxylate

  • MDL number MFCD12563836
  • Molecular Weight : 354.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1974

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1974
Availability details for
Quantity
  • Availability
  • Add to Cart