COM448686931 Combi-Blocks, Inc.

(4-Bromo-1h-indol-3-yl)-acetic acid methyl ester

  • MDL number MFCD09880075
  • Cas Number : 89245-37-4
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1977

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1977
Availability details for
Quantity
  • Availability
  • Add to Cart