COM448686933 Combi-Blocks, Inc.

2,3-dimethyl 5-(4-nitrophenyl)methyl 4H,6H,7H-pyrazolo[1,5-a]pyrazine-2,3,5-tricarboxylate

  • MDL number MFCD27578571
  • Cas Number : 1820619-25-7
  • Molecular Weight : 418.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1980
Availability details for
Quantity
  • Availability
  • Add to Cart