COM448686940 Combi-Blocks, Inc.

Methyl (2r)-2-amino-5-methylhexanoate

  • MDL number MFCD24642497
  • Cas Number : 1820579-86-9
  • Molecular Weight : 159.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1989

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1989
Availability details for
Quantity
  • Availability
  • Add to Cart