COM448686946 Combi-Blocks, Inc.

methyl 2-fluoro-4-(4-formylphenyl)benzoate

  • MDL number MFCD27578579
  • Cas Number : 1820673-58-2
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1999

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1999
Availability details for
Quantity
  • Availability
  • Add to Cart