COM448686949 Combi-Blocks, Inc.

methyl 4-[3-(trifluoromethoxy)phenyl]benzoate

  • MDL number MFCD27578582
  • Cas Number : 1433879-01-6
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2003
Availability details for
Quantity
  • Availability
  • Add to Cart