COM448686950 Combi-Blocks, Inc.

methyl 6-(thiophen-3-yl)pyridine-2-carboxylate

  • MDL number MFCD27578583
  • Cas Number : 1820608-45-4
  • Molecular Weight : 219.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2004

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2004
Availability details for
Quantity
  • Availability
  • Add to Cart